Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Refrigerated Incubators, Extra Large Capacity, Caron Products

Supplier: Caron Products

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

7001-75-2 7001-75-1 7001-50-3 7001-50-1 7001-50-2 7001-75-3
10033-946EA 49431.55 USD InStock
10033-946 10033-944 10033-834 10033-942 10033-832 10033-830
Refrigerated Incubators, Extra Large Capacity, Caron Products
Incubators Cooled Incubators
Caron's extra large capacity Refrigerated Incubators precisely control temperature conditions.

These incubators serve a wide range of applications, such as laboratory refrigerators, pharmaceutical refrigerators for long term storage of drug substances, temperature critical processes, temperature conditioning, and more. Standard temperature conditions range from 5 to 70 °C (41 to 158 °F).

Units are energy efficient due to Caron’s unique earth-friendly design features that yield up to 71% utility cost savings. A 17.8 cm (7") color touchscreen is intuitive and features easy-to-read icons and a familiar, user-friendly display that can be operated with gloves on. All operating conditions can be viewed simultaneously, even from across the room.

Note: View the Product Accessories List, in Datasheets under Related Information. To order specific accessories, search by part number.

These stainless steel, two-door (50 cu.ft.) and three-door (75 cu.ft.) models are the ultimate choice for high volume, refrigerated storage.

Ordering information: The 1415.8 l (50 cu.ft.) units are supplied with eight perforated stainless steel shelves. The 2123.8 l (75 cu.ft.) units are supplied with twelve perforated stainless steel shelves.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: Caron provides customers with top-quality products backed by technical service and support for the life of the product.

Extended Warranty Packages offer peace of mind with up to an additional 1 year or 3 years on top of the original warranty, covering parts and labor for any manufacturer's issues that may occur during this time. Purchase of an Extended Warranty is only valid prior to the original warranty expiration and is only available for U.S. Customers. See table below for more information.

PM Service Kits help to minimize downtime by providing routine maintenance items for equipment. See table below for more information.

PM Service Kits for Accessories help to minimize downtime by providing routine maintenance items for accessories. See table below for more information.

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty