Would you like to log in again?
Incu-Shaker™ Incubators, Chemglass
Supplier: Chemglass
CLS-2543-0R2
10034-198EA 15899.8 USD InStock
10034-198
Incu-Shaker™ Incubators, Chemglass
Shakers and Mixers Shaking Incubators
The new Incu-Shaker™ series of shaking incubators provide digitally regulated temperature with precisely controlled circular (orbital) mixing, optimized for the culture of a wide variety of cell types
The standard platform (included with both models) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays. Convenient MAGic Clamp™ platforms, flask clamps and tube racks are also available for use with Erlenmeyer flasks and test tubes. This unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes. Each clamp utilizes a powerful (patent pending) magnetic base to securely attach the clamp to the shaking platform without the use of screws, bolts or tools.
Temperature, shaking speed and run time are displayed on the large LED control panel located on the front of the unit. The integral microprocessor includes a constant monitoring system that verifies and maintains accuracy through the duration of the program. Sophisticated over-temperature and over-speed controls ensure long life, safety and sample integrity.
With similar features to the Incu-Shaker™ Mini, the 10L Incu-Shakers™ are equipped with a large 18 x 18" platform for high capacity applications.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty