Would you like to log in again?
Powder Transfer Adapters, PTFE, Chemglass
Supplier: Chemglass
CG-1057-05 CG-1057-06 CG-1057-03 CG-1057-04 CG-1057-01 CG-1057-07
10034-274EA 78.72 USD InStock
10034-274 10034-276 10034-282 10034-284 10034-280 10034-278
Powder Transfer Adapters, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters simplify the transfer of powders/solids from flasks into vials.
  • Tapered interior of adapter prevents hold-up
  • Reduces steps by transferring directly to a vial, no other tools are needed
  • Allows transfer of samples without exposure to air or moisture
PTFE adapters have a knurled gripping ring at the top to facilitate removal of the adapter from the flask joint. Adapters are available to accommodate sample vials (GPI threads) and scintillation vials (22 mm threads). To use, screw the vial into the top of the transfer adapter. Insert the other end of the adapter into a flask joint. Invert assembly and gently tap vial on a soft surface to transfer contents from flask into the vial.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty