Would you like to log in again?
Tube Brushes, Flexible, Chemglass
Supplier: Chemglass
CG-1142-02 CG-1142-01
10034-288EA 22.19 USD InStock
10034-288 10034-290
Tube Brushes, Flexible, Chemglass
Brushes
Brushes feature polypropylene bristles attached to flexible glavanised steel wire.
  • Ideal for cleaning long tubular items
  • Fits tube with an I.D. of 0.5" to 1"
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty