Would you like to log in again?
Test Tube Brush, Chemglass
Supplier: Chemglass
CG-1143-01
10034-292EA 21.28 USD InStock
10034-292
Test Tube Brush, Chemglass
Brushes
This brush has polypropylene bristles and can be spread apart to clean tubes having an I.D. of 14 mm or greater.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty