Would you like to log in again?
Laboratory Cleaning Brushes, Chemglass
Supplier: Chemglass
CG-1144-01
10034-294EA 16.46 USD InStock
10034-294
Laboratory Cleaning Brushes, Chemglass
Brushes
These laboratory brushes feature polypropylene bristles with foam bristles at the tip.
Ideal for use with wide mouth bottles, graduated cylinders and beakers. Width of brush tip is 2".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty