Would you like to log in again?
Whatman™ Grade 597 and 597 ¹/₂ Qualitative Filter Papers
Supplier: Whatman products (Cytiva)
Whatman™
10311852 10311810 10311812 10311854 10311856 10311809 10311841 10311822 10311809 10311842 10311820 10311807 10311814 10311808 10311887 10311845 10311808 10311811 10311804 10311862 10311843 10311847 10311814 10311841 10311853 10311843 10311851 10311851 10311804 10311812 10311820 10311811 10311854 10311887 10311856 10311852 10311842 10311853 10311844 10311847 10311844 10311862 10311897 10311810 10311845 10311822 10311807 10311897
10034-682PK 463.5 USD InStock
10034-682 CA11012-690 14224-640 14224-442 14224-420 14224-440 10034-680 10034-668 89233-650 14224-424 CA11012-750 CA14224-434 CA11012-676 10034-664 CA11012-754 10034-666 11008-542 CA14224-436 CA11012-756 CA11008-542 CA11012-706 CA11008-544 CA14223-590 74330-452 CA14223-536 10034-670 10034-672 CA14224-440 CA11012-660 CA11012-682 14224-436 CA89233-650 10034-678 CA74330-452 CA14224-420 CA14224-442 CA14224-640 14223-590 14224-434 CA11012-744 10034-674 CA11012-688 CA14224-424 10034-676 74300-006 CA74300-006 11008-544 14223-536
Whatman™ Grade 597 and 597 ¹/₂ Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
  • Increased total loading capacity as more filter area is available
Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles. Cellulose materal (98% alpha cellulose content min.). Circles format.
A medium fast filter paper with medium to fine particle retention. Used for a wide variety of analytical routine applications in different industries like food testing (e.g., determination of fat content) or removal of carbon dioxide and turbidity from beverages (e.g., beer analysis).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty