Would you like to log in again?
Whatman™ Grade 0860 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10334551 10334553 10334553 10334547 10334547 10334607 10334551 10334607
89233-664PK 173.26 USD InStock
CA76423-760 CA89233-664 89233-664 CA11013-894 76423-760 10034-756 CA11013-262 10034-758
Whatman™ Grade 0860 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Comparable to Grade 0858 but with a smooth surface, slightly thinner and faster.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty