Would you like to log in again?
Whatman™ Standard Cellulose Extraction Thimbles, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10350274 10350240 10350215 10350234 10350242 10350211 10350211 10350275 10350265 10350227 10350238 10350241 10350225 10350223 10350306 10350215 10350255 10350252 10350225 10350227 10350247 10350261 10350324 10350273 10350275 10350324 10350238 10350273 10350217 10350243 10350252 10350274 10350220 10350437 10350236 10350250 10350245 10350437 10350236 10350287 10350219 10350261 10350219 10350234 10350226 10350223 10350247 10350245 10350226 10350250 10350306 10350255 10350265 10350287 10350243 10350220 10350241
28152-848PK 143.6 USD InStock
28152-848 89233-658 CA11013-558 28152-846 CA11013-556 28152-844 89233-656 10034-800 CA11013-550 10034-802 CA11013-448 10034-792 CA11013-844 CA89233-658 CA11013-842 10034-794 CA89233-656 CA11013-444 10034-790 CA11013-440 CA11013-560 10034-796 10034-798 CA28152-846 CA11013-834 74330-280 CA11013-536 10034-780 CA11013-458 10034-782 CA11013-532 CA11013-530 CA11013-450 10034-788 10034-820 10034-784 10034-786 10034-818 10034-816 10034-814 10035-876 10035-878 CA11013-528 CA11013-846 CA11013-426 CA11013-546 CA11013-540 CA11013-462 10034-812 10034-778 10034-810 CA11013-460 10034-808 10034-806 10034-804 CA11013-538 CA11013-416
Whatman™ Standard Cellulose Extraction Thimbles, Whatman products (Cytiva)
Extractors Extraction Thimbles
Grade 603 Standard Cellulose Extraction Thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty