Would you like to log in again?
Whatman™ Mixed Cellulose Ester Membranes, Non-Sterile, 0.2 µm, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Membra-Fil®
10401714 10401731 10401721 10401712 10401714 10401721 10401726 10401726 10401712 10401706 10401706 10401731
10034-924PK 184.3 USD InStock
CA74300-038 10034-924 74330-188 10034-922 CA74330-188 CA11012-764 10034-920 CA11013-806 CA11013-958 74300-036 74300-038 CA74300-036
Whatman™ Mixed Cellulose Ester Membranes, Non-Sterile, 0.2 µm, Whatman products (Cytiva)
Filters Membrane Filters
Whatman non-sterile mixed cellulose ester (MCE) filters from Cytiva are suitable for E. coli culturing, enumeration of bacteria, and other culturing and cell counting methods.
  • Plain and gridded options for ease of colony and cell counting methods
  • Supports visual enumeration of bacteria
  • Appropriate for bacterial culturing, including E. coli culturing
  • Biologically inert with good thermal stability
  • Uniform microporous structure results in high flow rates when used for filtering
Whatman MCE filters from Cytiva business are composed of cellulose acetate and cellulose nitrate, giving the membrane a more uniform surface than pure nitrocellulose filters. Browse Cytiva's other MCE filter options to find the right filter for your application.
MCE filters support microbiology applications such as E. coli culturing. The porous membrane structure enables the uptake of growth media to the bacterial colonies without chemically inhibiting their growth. The gridded surface simplifies visual enumeration of bacteria, including E. coli, enterococci, clostridia, fecal coliforms, staphylococci, and Pseudomonas aeruginosa.
Plain and gridded MCE membrane options are available. The gridded option has grid lines spaced at 3.1 mm intervals. Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty