Would you like to log in again?
Whatman™ Cellulose Acetate Membrane Filters
Supplier: Whatman products (Cytiva)
Whatman™
10404180 10404106 7000-0004 10404006 10404026 10404131 10404106 10404126 10404139 10404112 10404006 7000-0002 10404044 10404026 10404031 10404044 7000-0002 10404180 10404114 10404012 10403112 10404001 10404014 10404012 10403112 10404001 10404126 10404112 10403012 10404014 10404139 7001-0004 10404031 10404131 7001-0004 10404114 10403012 7000-0004
10035-528PK 209.95 USD InStock
10035-528 CA11012-650 CA11013-178 10034-944 CA11013-032 10034-942 CA74330-202 10034-940 CA74330-246 CA11012-776 CA74330-204 89014-302 CA74330-304 CA11012-904 10034-938 10034-936 CA11013-808 74330-198 CA11013-968 CA11013-824 CA11013-966 CA11013-962 CA74330-198 CA11012-740 10034-934 CA89014-302 10034-954 10034-932 CA11012-800 10034-950 74330-304 CA11012-738 74330-204 10034-948 74330-202 74330-246 10034-946 10035-740
Whatman™ Cellulose Acetate Membrane Filters
Filters Membrane Filters
Cytiva's Whatman cellulose acetate membrane filters have very low protein binding, which minimizes sample loss when filtering protein-based aqueous samples.
  • Pure cellulose acetate composition, minimizing sample contamination
  • Hydrophilic, for use with aqueous and alcoholic media
  • High solvent resistance, particularly to low molecular weight alcohols
  • High physical strength and a heat resistance up to 180 °C
  • Can be sterilized by most common methods without sacrificing membrane integrity
Due to their high chemical resistance, cellulose acetate membrane filters reliably prepare liquid scintillation cocktail for scintillation counter measurement.
High chemical resistance membranes.
Additionally, the highly heat resistant Whatman cellulose acetate membranes can filter hot liquids and hot gases.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty