Would you like to log in again?
Whatman™ Polyamide Membrane Circles, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10414112 10414006 10414014 10414012 10414114 10414014 10414106 10414114 10414012 10414106 10414112 10414006
28152-899PK 223.25 USD InStock
CA11013-038 28152-899 CA28152-892 CA11013-046 28152-896 CA28152-894 10035-004 28152-894 CA28152-896 10035-002 28152-892 CA28152-899
Whatman™ Polyamide Membrane Circles, Whatman products (Cytiva)
Filters Membrane Filters
Whatman polyamide membranes are made from pure polyamide making them the universal filter for clarification and sterile filtration.
Polyamide membrane filters are mechanically very strong and exhibit excellent wet strength and dry strength. They are hydrophilic making them suitable for aqueous and organic solutions. The membrane filters can be used up to 135 °C.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty