Would you like to log in again?
Whatman™ Syringe Filter Holder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
10461000 10461000
10035-078PK 244.93 USD InStock
CA11012-948 10035-078
Whatman™ Syringe Filter Holder, Whatman products (Cytiva)
Filter Holders
Syringe Filter Holder made of polysulfone is useful in clarification and sterile filtration of liquids and gases under pressure.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty