Would you like to log in again?
FILTER HOLDER050/0 PSU 50MM PK5
Supplier: Cytiva
Filter Holder, Polysulfone, Silicone O Ring, 50mm, 5/pk^ Has a luer lock fitting( female) on one side, and a barb fitting on the other side.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
FILTER HOLDER050/0 PSU 50MM PK5 10035-080 10461200 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty