Would you like to log in again?
Whatman™ Puradisc Aqua 30 mm Syringe Filter, 0.45 µm, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Puradisc™ Puradisc™
10462655 10462650 10462655 10462656 10462656 10462650
10035-110PK 188.69 USD InStock
CA11013-434 10035-110 CA11013-156 CA11013-130 10035-108 10035-106
Whatman™ Puradisc Aqua 30 mm Syringe Filter, 0.45 µm, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Puradisc Aqua 30 from Cytiva's business is specifically designed for environmental sample filtration prior to dissolved organic carbon (DOC) and chemical oxygen demand (COD) analysis.
  • From the Puradisc FP 30 range
  • Low protein binding, hydrophilic cellulose acetate syringe filter media
  • Prewashed cellulose acetate (CA) syringe filter for reduced organic carbon extractables
  • Designed for sample preparation in support of dissolved organic carbon and chemical oxygen demand analysis
  • Standard inlet and outlet luer connectors
DOC (dissolved organic carbon) and COD (chemical oxygen demand) analysis are common methods to determine organic pollutant levels.
Precisely filtering a sample is a crucial step in DOC and COD, and the quality of the syringe filter should preserve sample integrity. Aqua 30 Puradisc syringe filters come prewashed, designed to minimize carbon leaching from the cellulose acetate membrane that could otherwise lead to sample contamination.
Water and Waste GmbH, a premier Austrian water-testing laboratory has high praise for Cytiva's Whatman syringes and membrane filters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty