Would you like to log in again?
Whatman™ Polydisc GW In-Line Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Polydisc™
10463400 10463401 10463400 10463401
10035-122PK 305.9 USD InStock
10035-122 10035-124 CA11012-972 CA11013-490
Whatman™ Polydisc GW In-Line Filters, Whatman products (Cytiva)
Filters In-Line Filters
Cytiva's Polydisc GW (Ground Water) is an in-line filter specifically designed for preparation of ground water samples for dissolved heavy metal analysis.
  • Hydrophilic, durable nylon membrane is compatible with aqueous samples
  • 100% quartz fiber prefilter with low heavy metal content allows filtration of larger volumes of even heavily particle-laden samples
  • Certified low background metal values for heavy metal testing
  • Large 20.4 cm² filtration area
  • Tubing nozzle compatible with 6 - 14 mm ID hose
Polydisc GW’s large filter surface, durable nylon Whatman membrane, and quartz fiber prefilter result in high dirt-loading capacity. The filter was designed in line with regulatory requirements for groundwater testing found in US EPA Method 3005A.
Heavy metal testing in ground water often requires an aqueous filter to reduce a sample’s turbidity, but results can be skewed if the filter itself contains heavy metals. Cytiva certifies each Polydisc GW batch for low background values for the determination of trace elements.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty