Would you like to log in again?
Whatman™ ReZist Syringe Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
ReZist™ Whatman™ ReZist™
10463523 10463503 10463513 10463523 10463525 10463543 10463500 10463505 10463703 10463515 10463713 10463545 10463703 10463535 10463535 10463513 10463515 10463543 10463505 10463503 10463545 10463525 10463500 10463533 10463713 10463533
14224-464PK 355.29 USD InStock
14224-464 14224-462 CA11013-496 CA11012-970 14224-466 CA11013-194 CA11013-192 CA11012-976 10035-150 10035-172 10035-154 10035-152 10035-158 10035-156 CA11013-504 CA14224-462 CA11013-168 CA11013-486 CA11013-200 CA14224-466 CA11013-164 CA14224-464 10035-162 10035-160 10035-166 10035-164
Whatman™ ReZist Syringe Filters, Whatman products (Cytiva)
Filters Syringe Filters
Whatman ReZist range of syringe filters has been specifically designed to be resistant to organic solvents.
  • Integral, permanently hydrophobic PTFE membranes
  • Polypropylene support
  • Extremely high chemical resistance
  • Recyclable after use
These filters are highly suitable for the clarification of aggressive organic solvents. ReZist 30 mm filters can also be used as a venting filter for small vessels.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty