Would you like to log in again?
Whatman™ ReZist Air Venting Filter, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
ReZist™ Whatman™ ReZist™
10463607 10463607 10463609 10463608 10463609 10463608
10035-170PK 653.6 USD InStock
CA76423-782 10035-170 CA11013-166 76423-782 10035-168 CA11012-966
Whatman™ ReZist Air Venting Filter, Whatman products (Cytiva)
Filters In-Line Filters
ReZist in-line filters are lightweight air venting units suitable for protective vents, in-line filtration, and isolation applications when working with chemically harsh solvents.
  • Integral hydrophobic PTFE venting membrane is resistant to harsh solvents
  • Can be autoclaved multiple times without loss of integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight: avoids the collapsing of tubing that can be caused by heavy filter devices
  • Appropriate for in-line HPLC sample preparation
This device features a PTFE venting membrane, which is well suited for chemically aggressive solutions, reagents, and organic solvents.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty