Would you like to log in again?
Whatman™ Grade 93 Qualitative Filter Papers
Supplier: Whatman products (Cytiva)
Whatman™
1093-6215 1093-111 1093-110 1093-935 1093-111 1093-126 1093-126 1093-125 1093-6215 1093-930 1093-125 1093-930 1093-110 1093-935
10035-714PK 18.29 USD InStock
CA11013-338 CA28449-026 CA28449-048 CA11013-312 10035-714 CA11013-340 CA11013-284 28449-026 28449-048 CA11013-290 10035-242 10035-250 10035-248 10035-246
Whatman™ Grade 93 Qualitative Filter Papers
Filters Filter Paper Qualitative Filter Paper
These cellulose filter papers are intermediate in speed and retention between grades 1 and 4.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate. The resins do, however, contain nitrogen so these grades should not be used in Kjeldahl estimations, etc.
Available in a dispenser pack, which can be attached to the wall or bench, placed on a shelf either upright or flat, and used as a normal carton or as a convenient dispenser. The envelopes are released individually for easy one-at-a-time removal.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty