Would you like to log in again?
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 41, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1441-042 1441-125 1441-125 1441-050 14418296 1441-060 1441-047 1441-185 1441-150 1441-090 10380204 1441-240 10380205 1441-240 1441-060 1441-110 1441-042 1441-055 1441-050 10380204 1441-110 1441-070 1441-070 1441-917 1441-185 10380206 10380206 1441-866 10380205 1441-055 1441-320 1441-6309 1441-917 14418296 1441-866 1441-6309 1441-320 1441-047 1441-150 1441-090
28478-080PK 124.26 USD InStock
CA11013-754 28478-080 CA10147-984 28477-974 28477-994 CA28297-291 CA76292-864 76423-770 CA28297-295 28478-068 CA76292-866 28478-046 28478-002 CA28478-110 28478-024 CA76292-868 28478-104 CA28478-080 CA11013-642 10147-984 CA11013-640 CA11013-360 CA28477-994 10035-262 28297-291 76292-864 CA28477-974L CA28478-104 10035-264 76292-866 CA76423-770 28478-110 76292-868 28297-295 10035-266 CA28478-068 CA28478-024 CA28478-046 CA28478-002 10035-268
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 41, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 41 environmental pollution papers are among the fastest filter papers in the Whatman ashless range. This gravimetric filter is well suited for quantitative analysis of soil and air.
  • Extremely low ash content (0.007% ash maximum)
  • 20 µm nominal particle retention rating
  • Fast filtration speed (Herzberg 54 s)
  • Commonly used for routine air pollution analysis and soil pollution analysis
Whatman Grade 41 filters from Cytiva's business are suitable for quantitative air pollution analysis as a paper tape for impregnation when determining gaseous compounds at high flow rates.
Fast ashless paper for pollution analysis. Grade 41 filters are also commonly used in preparing solid environmental samples (e.g., soils, sludges) for metals analysis after nitric acid digestion. Grade 41 is available in a convenient, disposable, 47 mm filter funnel with a 250 ml capacity. The 47 mm diameter filter can be easily removed for further analysis or culturing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty