These borosilicate glass filters are widely used as total suspended solids filters for applications testing both potable water and waste water. GF/C can also function as a filter for liquid scintillation counting, cell harvesting, industrial waste analysis and binding assays that require high loading capacity.
- Fine particle retention
- Good flow rate and loading capacity
- Binder free