Would you like to log in again?
Whatman™ 1PS Phase Separator Papers
Supplier: Whatman products (Cytiva)
Whatman™
2200-240 2200-270 2200-270 2200-150 2200-070 2200-110 2200-090 2200-110 2200-240 2200-150 2200-185 2200-090 2200-125 2200-070 2200-125 2200-185
28491-204PK 232.11 USD InStock
28491-204 28491-226 CA28491-168 CA28491-124 CA28491-102 CA28491-146 28491-102 28491-124 28491-168 28491-146 CA11013-064 CA28491-180 28491-180 10035-338 CA28491-226 CA28491-204
Whatman™ 1PS Phase Separator Papers
Filters Filter Paper Phase Separation Paper
Whatman 1PS Phase Separator from Cytiva's business is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
  • Impregnated with stabilized silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Solvent extraction can be time-consuming and messy when numerous separations are required
Mixed phases are poured directly into the Whatman 1PS Phase Separator filter. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
Cytiva's Whatman 1PS Phase Separator is impregnated with stabilized silicone, creating a hydrophobic filter paper that can replace separatory funnel use in certain applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty