Would you like to log in again?
Whatman™ Polycap™ HD Disposable Filter Capsules
Supplier: Whatman products (Cytiva)
Whatman™ Polycap™ Polycap™
6703-3621 2713 2610T 2611T 2713 2610T 6703-7550 2814T 6703-3650 6703-3610 6703-3611 2712T 6703-7550 2813 2812T 6703-7521 6703-7511 6703-3610 2713T 2613T 2711T 2813 2612T 2714T 2710 2711T 6703-3621 6703-3611 2812T 2611T 2612T 2814T 2813T 2813T 2712M 6703-3650 2810T 2810T 2614T 2614T 6703-7511 2712M 6703-7510 2710 2714T 6703-7510 2611 6703-7521 2611 2713T 2712T 2613T
28137-894EA 77.9 USD InStock
28137-894 28137-896 28137-890 CA11013-232 28137-892 CA28137-900 CA28137-902 CA11012-832 28137-898 10034-564 CA11012-834 CA11012-868 10035-776 10035-414 10035-772 CA11013-004 10035-418 CA11013-244 28137-902 28137-900 CA11012-844 10035-384 10035-386 10035-380 10035-382 10035-426 10035-420 10035-388 CA11013-714 CA11013-732 10035-826 CA28137-888 CA11012-852 CA11013-092 CA11013-090 10035-832 10035-834 CA11013-726 CA11013-724 10035-830 CA11013-228 CA28137-892 CA11013-700 CA28137-894 CA11013-740 CA28137-890 CA11012-862 CA28137-896 28137-888 CA28137-898 10035-360 10035-768
Whatman™ Polycap™ HD Disposable Filter Capsules
Filters Capsule Filters
Polycap HD (Heavy Duty) capsule filter offers a broad range of particle size retention ratings and excellent filtrate purity with polypropylene filters (PP).
  • Polypropylene filters support use with a broad range of solutions, pH solutions, and temperatures
  • Sturdy construction leads to high flow and high retention capacity
  • Can be autoclaved with steam at 121 °C for 20 minutes
  • Manual vent with luer lock to bleed air from upstream or serve as an injection or sample port
Chemical processing applications often require a capsule filter to ensure a finishing step yielding high purity final product. Different solution temperatures and pH values can complicate filtration unless an adequate filter capsule is selected.
Due to its heat sealed construction and PP filter, Polycap HD (Heavy Duty) is suitable for filtering chemical solutions across wide temperature and pH ranges.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty