Cytiva's Whatman Polycap TF filters are made with durable, hydrophobic polytetrafluoroethylene (PTFE) membranes in a polypropylene housing for use with organic solvents and chemically aggressive solutions.
- PTFE filter membrane is resistant to most solvents
- Available in 0.05, 0.1, 0.2, 0.45, and 1.0 µm pore sizes
- 0.05 µm pore size capsules capsules designed for ultra-clean applications
- 1.0 µm pore size capsules used for extended life and filtration of highly contaminated solutions
- Autoclavable capsule filtration device if sterility is required
Filtration of organic solvents and aggressive solutions requires a durable membrane for filtration without sample contamination.
Autoclavable Polycap TF capsule filters feature PTFE membranes with high chemical resistance. Common applications include venting, in-line filtration to sterilize and remove particles from air/gas/liquid streams, filtration to ensure clean inert gases in vial filling lines, and isolation of exhaust from fermenters.
Polycap TF filters support use in the electronics, pharmaceutical, and biopharmaceutical industries, because they are photoresistant and can be used to vent and perform in-line filtration for production systems in biological environments.
Certifications: Manufactured according to ISO9001:2008 QMS standards