Would you like to log in again?
Whatman™ Polycap™ AS Disposable Filter Capsules, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Polycap™ Polycap™
6708-3602 6706-3602 6705-3602 2707NS 6709-3602 6706-3602 6708-3602 2606T 6705-3604 2707NS 2606T 6709-3602 2706T 2706T
28137-698EA 80.57 USD InStock
28137-698 CA28137-690 28137-688 28137-700 CA28137-698 CA28137-700 CA11012-866 CA11012-836 28137-908 10035-358 28137-690 CA11013-706 10035-376 10035-378
Whatman™ Polycap™ AS Disposable Filter Capsules, Whatman products (Cytiva)
Filters Capsule Filters
Cytiva's Whatman Polycap AS (Aqueous Solution) combines a nylon membrane filter and glass microfiber (GMF) prefilter for a long lifetime device that can filter large volumes of even difficult, particulate-laden samples.
  • First layer (GMF) acts as a prefilter to ensure longer membrane (0.2, 0.45, and 1.0 µm) life and higher filtration efficiency
  • Nylon membrane filter layer is inherently hydrophilic, has low extractables, is biosafe, and has excellent flow rates
  • Polypropylene housing is thermally fused (no glues, adhesives, or extraneous materials), reducing extractable levels
  • Can be tested with bubble point, pressure decay, or forward flow methods
  • Available in presterilized and nonsterile (autoclavable) versions
Polycap AS are Whatman capsule filters specifically designed for high-volume filtration of aqueous sample solutions in a compact size.
Certifications: Manufactured according to ISO9001:2008 QMS standards
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty