Would you like to log in again?
Whatman™ Polycap™ GW, Ground Water Sampling Capsule Filter, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™ Polycap™ Whatman™ Polycap™
6724-6004 6714-6004 6714-6004 6724-6004
10035-490PK 7058.5 USD InStock
CA11013-920 CA13503-498 10035-490 13503-498
Whatman™ Polycap™ GW, Ground Water Sampling Capsule Filter, Whatman products (Cytiva)
Filters Capsule Filters
Specifically designed for field sampling, Cytiva's Whatman Polycap GW ground water sampling filter capsules are a polyethersulfone (PES) filter that can be used as a convenient in-line filter unit.
  • Stepped hose barb fittings allow for connection directly to outlet of a sampling pump with various size tubing
  • Effective filtration surface area of 600 cm² to ensure rapid sample collection
  • PES filters in thermally fused housing eliminate sample contamination that could come from glues, adhesives, metals, epoxies, or extraneous materials
  • Lot number printed on each unit for traceability supports use in regulated field sampling methods such as EPA Method 3005 for ground water analysis
  • Well suited for ground water sampling as a dissolved metals filter
The United States Environmental Protection Agency (EPA) and other environmental protection agencies have protocols specifying support of ground water sampling with a 0.45 µm filter prior to analyzing dissolved or suspended metals.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty