Would you like to log in again?
Whatman™ Grade GF 6 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10370007 10370005 10370003 10370008 10370012 10370002 10370005 10370002 10370010 10370008 10370018 10370018 10370003 10370012 10370050 10370019 10370010 10370020 10370007 10370006 10370019 10370004 10370011 10370050 10370004 10370006 10370011 10370020
10035-670PK 529.15 USD InStock
CA11013-534 CA11013-852 CA11013-554 CA11013-850 CA11013-454 CA11013-552 10035-670 10035-672 10035-678 10035-674 CA11013-428 10035-676 CA11013-446 CA11013-544 CA11013-564 CA11013-442 10034-832 10034-830 10035-680 10035-880 10035-682 10035-666 10035-668 CA11013-838 10035-684 CA11013-836 10035-664 CA11013-856
Whatman™ Grade GF 6 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman Grade GF 6 glass microfiber filters from Cytiva's business are suitable for water and wastewater analysis, with extremely fine particle retention and added strength.
  • Sub micron filter retention efficiency to capture extremely small particles
  • Inorganic binder for increased strength
  • Temperature-resistant up to 500 °C
  • Can be used as a pre-filter for dense samples
  • Frequently used in waste water analysis
Grade GF 6 glass filters are suitable for water pollution and waste water analysis. Both require a sub micron filter that can capture tiny particles without weakening. Other applications include: Use as a beer filter in applications including protein removal; Determination of chlorophyll and phytoplankton residue; Determination of filterable substances and residue on ignition; Analysis of aggressive media (e.g., acidic gases); Determination of iron content in the presence of iron oxides; Liquid scintillation measurement.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty