Would you like to log in again?
Whatman™ Sterile Cellulose Nitrate Membrane Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10401170
10035-698PK 191.48 USD InStock
10035-698
Whatman™ Sterile Cellulose Nitrate Membrane Filters, Whatman products (Cytiva)
Filters Membrane Filters
Recommended for microbiological culturing and sample preparation, these cellulose nitrate membrane filters offer very narrow pore size distribution and low levels of extractables.
  • Sterile membrane filters eliminate the risk of contamination that could impact analysis
  • High strength and flexibility
The cellulose nitrate sterile membrane filters are well suited for separation applications involving particles up to 12 µm. After sample preparation, filtration residue is almost completely retained by the plain white membrane surface, supporting clear microscopy and physical recovery of deposits.
Most membranes are inherently brittle and difficult to handle; some may be damaged as they are loaded into holders or during standard use. Whatman cellulose nitrate membrane filters are designed to tolerate rough handling.
Pharmaceutical, immunological, biomedical tissue culture, and trace analysis applications rely on knowing that filtration preserves the integrity of the original sample. Because Whatman cellulose nitrate membrane filters have a low level of extractables, they are a good choice for sample preparation of critical samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty