Would you like to log in again?
Whatman™ Vacuum-Type Glass Membrane Holder, Glass Reservoir, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Whatman™
1961-054 1961-054
10035-722PK 193.39 USD InStock
CA11013-188 10035-722
Whatman™ Vacuum-Type Glass Membrane Holder, Glass Reservoir, Whatman products (Cytiva)
Filter Holders
Produced from borosilicate glass and available with a choice of support screen.
  • Suitable for aqueous and organic solvent filtration
The funnel seal ensures that the sample does not bypass the membrane and that particulates are retained on the surface of the membrane. The sintered glass support is recommended for filtration and biological analysis. The 304-stainless steel support screen is suitable for use with proteinaceous solutions.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty