Would you like to log in again?
Whatman™ Anotop HPLC Syringe Filters, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Anotop™ Whatman™ Anotop™
2001-0100 2002-5100 2002-5200 2001-0200 2002-5100 2002-5200 2001-0100 2001-0200
10035-726PK 928.15 USD InStock
10035-726 CA11013-058 10035-728 CA11013-056 10035-732 CA11013-054 10035-730 CA11013-060
Whatman™ Anotop HPLC Syringe Filters, Whatman products (Cytiva)
Filters Syringe Filters
Whatman Anotop HPLC syringe filter products from Cytiva's business are specifically designed for effective UHPLC and HPLC sample preparation.
  • Syringe filter removes particulates that could cause damage, supporting longer column life
  • Low levels of UV-absorbing extractable substances for consistent HPLC sample preparation
  • Anopore inorganic aluminum membrane filters a broad range of solvents
  • High pore-size precision due to capillary pore structure
Cytiva's Anotop HPLC Syringe Filter helps isolate peaks of interest by filtering particles that could otherwise interfere with accurate analysis prior to liquid chromatography. Anotop LC’s pigment-free polypropylene housing and Anopore inorganic filter media provide low levels of UV-absorbing extractables and compatibility with many chemical solvents.
The Anotop aluminum-based Anopore membrane is effective in most organic solvents and aqueous samples. Made from inorganic materials, Cytiva's Anopore membranes are designed for chemical inertness and functionality in a wide range of temperatures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty