Would you like to log in again?
POLYDISC WPE 50 PK50 A/A
Supplier: Cytiva
In-line Filters - Polydisc WPE 50mm^ With 50um pore size screen. Inlet and Outlet are 1/8 inch NPT.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POLYDISC WPE 50 PK50 A/A 10035-762 2233 Pack of 50 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty