Would you like to log in again?
POLYCAP 36 1.0 PTFE A/A PK5
Supplier: Cytiva
Polycap* TF 36, 1.0 um, with SB inlet and outlet, 5/pk^ Capsules are made with durable, hydrophobic (PTFE) membranes in a polypropylene housing and are designed for use with organic solvents and chemically aggressive solutions.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POLYCAP 36 1.0 PTFE A/A PK5 10035-766 2603 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty