Would you like to log in again?
POLYCAP 36 0.2 PES A/A S PK5
Supplier: Cytiva
Polycap* TC (PES) 36, 0.2um, sterile, with SB inlet and outlet, 5/pk^ Are disposable, dual layer polyethersulfone (PES) membrane filtration capsules that provide efficient filtration for critical aqueous solutions.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
POLYCAP 36 0.2 PES A/A S PK5 10035-770 2642 Pack of 5 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty