Would you like to log in again?
Whatman™ Mini-UniPrep G2 Syringeless Filters
Supplier: Whatman products (Cytiva)
Whatman™ Mini-UniPrep™ G2
GN203NPUGMF GS503NPUAQU GN503NPEAQU GN203NPENYL GS203NPEORG GN203NPUORGSP GN203NPURC GN203NPEAQU GN203APEAQU GS203NPUGMFSP GN203APEAQU GN503NPUAQU GS203NPEAQUSP GN203NPERCSP GN503NPERC GN503NPUORG GN203NPENYLSP GN203NPURCSP GS203NPUAQUSP GS203NPEPP GN203NPEPPSP GN203NPENYLSP GN203APEORG GN203NPEAQUSP GN203NPUAQU GN203NPUAQUSP GN203NPEORGSP GN503NPUGMF GS203NPUGMF GS203NPUORG GN203NPUORG GN203NPUGMF GS203NPEORGSP GN503NPUORG GN203NPURCSP GN503NPURC GS203NPEPPSP GN203NPUAQUSP GN503NPUGMF GS203NPENYL GS203NPUGMF GN203NPERC GS503NPEAQU GN503NPEPP GN503NPURC GS203NPENYLSP GN503NPERC GN503NPEAQU GS503NPENYL GS503NPENYL GS203NPEPPSP GN203NPENYL GS203NPEORG GN203APEORG GN203NPUAQU GN203NPERCSP GS203NPEAQU GS203NPUAQU GN203NPUORGSP GS203NPEAQUSP GS503NPUORG GS503NPUORG GN203NPUGMFSP GN203NPEAQUSP GS203NPUORG GS203NPUORGSP GS203NPUORGSP GS503NPEAQU GN203NPEPP GS203NPUAQU GN203NPEORG GN203APEORGSP GN503NPEORG GN503NPEPP GN203NPURC GN203APEAQUSP GN203APEORGSP GS203NPUGMFSP GN203NPEPPSP GN203NPUGMFSP GS503NPEORG GN203NPEORGSP GN203NPERC GN203NPUORG GS203NPEORGSP GN503NPUAQU GN203NPEORG GN503NPENYL GS503NPEORG GN203NPEAQU GN503NPEORG GS203NPEAQU GS503NPUAQU GS203NPENYL GN203NPEPP GN203APEAQUSP GS203NPENYLSP GS203NPEPP GS203NPUAQUSP GN503NPENYL
10035-928PK 6326.63 USD InStock
10035-928 10035-924 10035-926 89234-982 10035-930 89234-980 10035-932 10035-938 10035-934 10035-936 10035-942 10035-944 10035-940 10035-946 10035-948 10035-952 10035-954 10035-950 10035-956 10035-958 CA10054-440 CA10054-442 10035-964 CA10054-444 CA10054-446 10035-966 10035-960 CA10054-448 10035-962 10035-968 CA10054-390 CA10054-392 CA89234-954 CA10054-394 CA10054-396 CA10054-398 10035-974 CA10054-432 CA10054-434 CA10054-436 10035-970 CA10054-438 10035-972 CA10054-418 CA89234-956 CA89234-958 CA10054-380 CA89234-962 89234-954 CA10054-382 CA89234-964 89234-956 CA10054-384 CA89234-960 CA10054-386 CA10054-420 CA10054-388 CA10054-422 CA10054-424 CA10054-408 10036-032 10036-030 10036-036 10036-034 CA89234-966 89234-958 CA89234-968 10036-038 89234-964 CA89234-974 89234-966 CA89234-976 89234-960 CA89234-970 CA10054-374 89234-962 CA10054-376 CA89234-972 CA10054-410 CA10054-378 CA10054-412 CA10054-414 CA10054-416 10036-040 10036-042 89234-968 CA89234-978 89234-976 89234-978 CA89234-980 89234-972 CA89234-982 89234-974 10035-920 CA10054-400 CA10054-402 89234-970 10035-922 CA10054-404 CA10054-406
Whatman™ Mini-UniPrep G2 Syringeless Filters
Filters Syringeless Filters
Mini-UniPrep G2 is an all-in-one syringeless filter with a borosilicate glass chamber. It replaces syringe filters for autosampler-supported UHPLC/HPLC sample preparation with easier handling and fewer consumable components.
  • Features an integrated autosampler vial, plunger with attached filter membrane, and septum/cap
  • Borosilicate glass vials eliminate plastic-based leachables that can originate from polypropylene vials
  • Designed to hold up to 0.5 ml of unfiltered sample that can then be loaded directly into an autosampler
  • Easy to see when a sample has been filtered
  • Wide range of membranes with 0.2 and 0.45 µm pore sizes to meet specific requirements
  • Also available in amber glass for light-sensitive samples
A variety of filter membrane media are available to meet your needs.
Mini-UniPrep Syringeless Filters combine a membrane filter, autosampler vial, septum, and plunger into a single device to provide efficient sample filtration for Ultra High Performance Liquid Chromatography (UHPLC)/High Performance Liquid Chromatography (HPLC) analysis.
Once compressed, Mini-Uniprep G2 can be placed into any autosampler that accepts a 12 mm × 32 autosampler vial for injection into your instrument. Alternatively, the septum can be pierced with a needle and the sample drawn off for manual injection into an analyzer.
Simplifying UHPLC/HPLC sample preparation minimizes instrument downtime resulting from use of unfiltered samples.
Browse the Whatman line to find the right filter and compressor for your application.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty