Would you like to log in again?
NA ELUANT PH 2.70 AA ANALYSIS 950ML CS4
Supplier: PICKERING LABS MS
Sodium Eluent, pH 2.70, 4 x 950 mL, For aminoacid analysis, should be protected from air to prevent contamination, buffers for hydrolysates
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
NA ELUANT PH 2.70 AA ANALYSIS 950ML CS4 100358-921 NA270 Case of 4 (950ml) Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty