Would you like to log in again?
BioFX® TMB One Component HRP Microwell Substrate, Surmodics
Supplier: Surmodics
BioFX®
TMBW-1000-01 TMBW-0100-01
100359-156EA 325 USD InStock
100359-156 100359-154
BioFX® TMB One Component HRP Microwell Substrate, Surmodics
Assays ELISAs
The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.
  • Prolonged Stability
  • Increased Sensitivity
  • Negligble backgrounds
  • Ease of Manufacturing
BioFX™ TMB One Component HRP Microwell Substrate gives assay developers the best possible tool to maximize shelf life stability and lower backgrounds while setting the industry standard for signal generation of the standard curve.
Certifications: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty