Would you like to log in again?
BioFX® TMB Super Sensitive One Component HRP Microwell Substrate, Surmodics
Supplier: Surmodics
BioFX®
TMBS-1000-01 TMBS-0100-01
100359-174EA 50 USD InStock
100359-174 100359-176
BioFX® TMB Super Sensitive One Component HRP Microwell Substrate, Surmodics
Assays ELISAs
The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution. Peroxidase reacts with the TMB substrate solution to produce a soluble blue reaction product that can be read at 370 nm or 620-650 nm for ELISA applications. Stop solutions can be added to prevent further color development and reading the absorbance values at the desired wavelengths.
  • Prolonged Stability
  • Increased Sensitivity
  • Negligible Backgrounds
  • Ease of Manufacturing
BioFX™ TMB Super Sensitive One Component HRP Microwell Substrate gives assay developers maximal signal generation and low backgrounds while maintaining a long shelf life.
Certifications: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty