Would you like to log in again?
BioFX® TMB One Component HRP Membrane Substrate, Surmodics
Supplier: Surmodics
BioFX®
TMBM-0100-01 TMBM-1000-01
100359-196EA 48 USD InStock
100359-196 100359-198
BioFX® TMB One Component HRP Membrane Substrate, Surmodics
Assays ELISAs
The one component 3,3’,5,5’-tetramethylbenzidine (TMB) substrate is supplied in a ready-to-use solution for membrane applications. The staining pattern provides a dark blue precipitate with low background to increase the signal-to-noise ratios in western blot, dot blots and line assays.
  • Intense staining pattern with low backgrounds
  • Room temperature stability for 2 years
  • Stable at 2-8° C for 4 years
BioFX® TMB One Component HRP Membrane Substrate provides increased sensitivity with low backgrounds.
Certifications: ISO 13485:2016 & 9001:2015 Certified
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty