Would you like to log in again?
BioFX® DAB Immunohistochemistry Substrate, Surmodics
Supplier: Surmodics
BioFX®
DABI-0100-2C
100359-228EA 120 USD InStock
100359-228
BioFX® DAB Immunohistochemistry Substrate, Surmodics
Assays Immunological Assays
DAB (3,3’-diaminobenzidine) is oxidized in the presence of peroxidase and hydrogen peroxide resulting in the deposition of a brown, alcohol-insoluble precipitate at the site of enzymatic activity.
  • Three year shelf life
  • Rapid and simple detection of peroxidase in immunohistochemical and in situ hybridization procedures
BioFX® DAB Immunohistochemistry Substrate is a two-component substrate system.
Certifications: ISO 13485:2016 and 9001:2015 certified.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty