Would you like to log in again?
ICP-MS Quality Control Standards for EPA Method 200.7, Inorganic Ventures
Supplier: Inorganic Ventures
IV-26-125ML IV-21-125ML IV-19-125ML IV-7-125ML
100359-856EA 403.76 USD InStock
100359-856 101372-472 100360-682 100359-528
ICP-MS Quality Control Standards for EPA Method 200.7, Inorganic Ventures
Standards Cationic Standards Multi-Element Standards
Quality Control Standard 26 is a certified reference material set in a nitric acid/hydrofluoric acid matrix for stability.
Standards for method 200.7 are designed for use with ICP-OES.
Certifications: ISO 17025 and ISO 17034 certified.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty