Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Picric Acid-Acetone for Brown & Brinn

Supplier: Ricca Chemical

R5861100-4A R5861100-500A
10036-328EA 175.91 USD InStock
10036-328 10036-326
Picric Acid-Acetone for Brown & Brinn
Picric Acid-Acetone for Brown & Brinn
Acetone 67-64-1 ACS (99.87%)
Picric Acid 88-89-1 ACS (0.09%)
Water 7732-18-5 ACS (0.04%)
Test Specification Result : Appearance Yellow liquid
Volumetric glassware complies with Class A tolerance requirements of ASTM E 288 and NIST Circular 434; it is calibrated before first use and
recalibrated regularly in accordance with ASTM E 542 and NIST Procedure NBSIR 74-461. Balances are calibrated regularly with weights certified
traceable to the NIST national mass standard. Thermometers and temperature probes are calibrated before first use and recalibrated regularly with a thermometer traceable to NIST standards. All products are prepared according to master documents that assure manufacture according to validatedmethods. Batch records document raw material traceability and production and testing history for each lot manufactured.


Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty