Would you like to log in again?
Magnesium ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGMG10-125ML CGMG1-500ML CGMG10-500ML
100360-618EA 148.35 USD InStock
100360-618 100360-032 100360-034
Magnesium ICP Standard, Inorganic Ventures
Standards Cationic Standards Magnesium Standards
Certified reference material for Magnesium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty