Would you like to log in again?
Silicon ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGSI10-125ML CGSI1-500ML CGSINA1-500ML CGSINA1-125ML CGSI10-500ML
100360-648EA 147.05 USD InStock
100360-648 100360-094 100360-096 100360-100 100360-300
Silicon ICP Standard, Inorganic Ventures
Standards Cationic Standards Silicium Standards
Certified reference material for Silicon, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty