Would you like to log in again?
Zirconium ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGZR1-500ML CGZR10-125ML CGZR10-500ML
100360-218EA 370.47 USD InStock
100360-218 100360-216 100360-678
Zirconium ICP Standard, Inorganic Ventures
Standards Cationic Standards Zirconium Standards
Certified reference material for Zirconium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty