Would you like to log in again?
Bismuth ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGBI10-125ML CGBI10-500ML CGBI1-500ML
100359-910EA 236.45 USD InStock
100359-910 100359-908 100360-586
Bismuth ICP Standard, Inorganic Ventures
Standards Cationic Standards Bismuth Standards
Certified reference material for Bismuth, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty