Would you like to log in again?
Gold ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGAU1-500ML CGAUN1-500ML
100360-604EA 504.7 USD InStock
100360-604 100360-602
Gold ICP Standard, Inorganic Ventures
Standards Cationic Standards Gold Standards
Certified reference material for Gold, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty