Would you like to log in again?
Phosphorus ICP Standard, Inorganic Ventures
Supplier: Inorganic Ventures
CGP1-500ML CGP10-125ML CGP10-500ML
100360-070EA 238.62 USD InStock
100360-070 100360-068 100360-630
Phosphorus ICP Standard, Inorganic Ventures
Standards Cationic Standards Phosphorus Standards
Certified reference material for Phosphorus, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty