Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Toyopearl® and TSK-GEL® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Supplier: Tosoh Bioscience
TSK-Gel®
43126 43127 16052 14479 21888 21448 07478 21887 14478 16050 14477 19802 43151 21449 43152 19818 07477 19026 43176 19956 43276 07476 43153 21301 43177 16173 14718 14720 44465 16172 43277 19955
100368-426EA 307.71 USD InStock
100368-426 101742-732 100368-260 100368-380 100367-978 100368-284 100368-286 101742-730 100367-718 100367-378 100367-872 100367-856 100367-614 100367-854 100367-656 100367-952 100368-428 100367-430 100367-450 100368-376 100368-170 100367-948 100367-628 100368-172 100367-726 100367-626 100368-094 100368-096 100368-330 100367-624 100367-380 100367-364
Toyopearl® and TSK-GEL® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Chromatography Sorbents HPLC Sorbents
Hydrophobic interaction chromatography resins can be applied to separate or purifiy proteins with similar chemical or structural properties, plasmids and monoclonal antibodies.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty