Would you like to log in again?
Toyopearl® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Supplier: Tosoh Bioscience
TSK-Gel®
43150 43125 43175 43278 19806
100367-312EA 832.21 USD InStock
100367-312 100367-532 100368-004 100368-168 100367-418
Toyopearl® Hydrophobic Interaction Chromatography Resins, Tosoh Bioscience
Chromatography Sorbents HPLC Sorbents
TOYOPEARL and TSKgel LabPak media products are small package sizes of TOYOPEARL and TSKgel bulk media products
The larger resin amounts in LabPak products allow the packing of wider bore and longer columns. This helps to accurately determine the resin's dynamic binding capacity, selectivity, column efficiency and operating conditions.
They contain three or four different ligand types offered for a particular chromatography mode.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty