Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Toyopearl® and TSK-GEL® LabPak Ion Exchange Chromatography Sorbents, Tosoh Bioscience
Supplier: Tosoh Bioscience
TSK-Gel®
43280 43380 43220 43210
102866-996EA 1537.4 USD InStock
102866-996 100367-422 100368-282 100368-174
Toyopearl® and TSK-GEL® LabPak Ion Exchange Chromatography Sorbents, Tosoh Bioscience
Chromatography Sorbents HPLC Sorbents
Toyopearl and TSK-GEL LabPak Media products are small package sizes of Toyopearl and TSK-GEL bulk media products.
The larger resin amounts in LabPak products allow the packing of wider bore and longer columns. This helps to accurately determine the resin's dynamic binding capacity, selectivity, column efficiency and operating conditions.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty